missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
| CAS | 10177-29-4 |
|---|---|
| Molecular Formula | C6H4ClNO2 |
| Molecular Weight (g/mol) | 157.55 |
| MDL Number | MFCD00128860 |
| InChI Key | IMRGVWZLCZERSQ-UHFFFAOYSA-N |
| Synonym | 4-chloronicotinic acid, 4-chloronicotinicacid, 4-chloro-3-pyridinecarboxylic acid, 4-chloro-nicotinic acid, 3-pyridinecarboxylic acid, 4-chloro, 3-carboxy-4-chloropyridine, 4-chlornicotinsaure, pubchem12989, acmc-1br3v, 4-chloropyridine-3-carboxyl |
| PubChem CID | 818229 |
| IUPAC Name | 4-chloropyridine-3-carboxylic acid |
| SMILES | OC(=O)C1=C(Cl)C=CN=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC441720050
|
N/A | 5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 10177-29-4 | |
| 157.55 | |
| IMRGVWZLCZERSQ-UHFFFAOYSA-N | |
| 818229 | |
| OC(=O)C1=C(Cl)C=CN=C1 |
| C6H4ClNO2 | |
| MFCD00128860 | |
| 4-chloronicotinic acid, 4-chloronicotinicacid, 4-chloro-3-pyridinecarboxylic acid, 4-chloro-nicotinic acid, 3-pyridinecarboxylic acid, 4-chloro, 3-carboxy-4-chloropyridine, 4-chlornicotinsaure, pubchem12989, acmc-1br3v, 4-chloropyridine-3-carboxyl | |
| 4-chloropyridine-3-carboxylic acid |
Specifications
| 10177-29-4 | |
| C6H4ClNO2 | |
| 4-chloronicotinic acid, 4-chloronicotinicacid, 4-chloro-3-pyridinecarboxylic acid, 4-chloro-nicotinic acid, 3-pyridinecarboxylic acid, 4-chloro, 3-carboxy-4-chloropyridine, 4-chlornicotinsaure, pubchem12989, acmc-1br3v, 4-chloropyridine-3-carboxyl | |
| OC(=O)C1=C(Cl)C=CN=C1 | |
| 157.55 | |
| 157.56 | |
| 4-Chloronicotinic acid |
| 95% min. (HPLC) | |
| MFCD00128860 | |
| IMRGVWZLCZERSQ-UHFFFAOYSA-N | |
| 4-chloropyridine-3-carboxylic acid | |
| 818229 | |
| 96% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
RUO – Research Use Only