accessibility menu, dialog, popup

UserName

NClOOH

CAS RN 10177-29-4

IUPAC Name: 4-chloropyridine-3-carboxylic acid
Synonyms: 4-chloropyridine-3-carboxylic acid
Molecular Weight (g/mol): 157.55
Molecular Formula: C6H4ClNO2
InChi Key: IMRGVWZLCZERSQ-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(Cl)C=CN=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results