missing translation for 'onlineSavingsMsg'
Learn More

4-Chloronicotinic acid, 96%

Catalog No. p-7047967
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2002903 1 g
AAL2002906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2002903 Supplier Thermo Scientific Chemicals Supplier No. L2002903

CAS: 10177-29-4 | C6H4ClNO2 | 157.55 g/mol

4-Chloropyridine-3-carboxylic acid is a building block used in a synthesis of benzocanthinones via tributyltin hydride radical induced cyclization. 4-Chloropyridine-3-carboxylic acid is also used in a synthesis of heterocyclic analogues of xanthone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloropyridine-3-carboxylic acid is a building block used in a synthesis of benzocanthinones via tributyltin hydride radical induced cyclization. 4-Chloropyridine-3-carboxylic acid is also used in a synthesis of heterocyclic analogues of xanthone.

Solubility
Soluble in organic solvents at 20 deg C

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers

Chemical Identifiers

CAS 10177-29-4
Molecular Formula C6H4ClNO2
Molecular Weight (g/mol) 157.55
MDL Number MFCD00128860
InChI Key IMRGVWZLCZERSQ-UHFFFAOYSA-N
Synonym 4-chloronicotinic acid, 4-chloronicotinicacid, 4-chloro-3-pyridinecarboxylic acid, 4-chloro-nicotinic acid, 3-pyridinecarboxylic acid, 4-chloro, 3-carboxy-4-chloropyridine, 4-chlornicotinsaure, pubchem12989, acmc-1br3v, 4-chloropyridine-3-carboxyl
PubChem CID 818229
IUPAC Name 4-chloropyridine-3-carboxylic acid
SMILES OC(=O)C1=C(Cl)C=CN=C1

Specifications

Melting Point ∽165°C (decomposition)
Quantity 1 g
Beilstein 9255849
Solubility Information Soluble in organic solvents at 20 deg C
Formula Weight 157.56
Percent Purity 97%
Chemical Name or Material 4-Chloronicotinic acid
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only