missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Chloromethyl)styrene (stabilized with TBC + ONP + 2-Nitro-p-cresol) 90.0+%, TCI America™
$93.21 - $503.30
Chemical Identifiers
| CAS | 1592-20-7 |
|---|---|
| Molecular Formula | C9H9Cl |
| Molecular Weight (g/mol) | 152.62 |
| MDL Number | MFCD00051362 |
| InChI Key | ZRZHXNCATOYMJH-UHFFFAOYSA-N |
| Synonym | 4-vinylbenzyl chloride, 1-chloromethyl-4-vinylbenzene, 4-chloromethylstyrene, 4-vinylbenzylchloride, benzene, 1-chloromethyl-4-ethenyl, 1-chloromethyl-4-ethenylbenzene, 4-chloromethyl styrene, p-vinylbenzyl chloride, unii-cjl4oj6bs0, cjl4oj6bs0 |
| PubChem CID | 74126 |
| IUPAC Name | 1-(chloromethyl)-4-ethenylbenzene |
| SMILES | ClCC1=CC=C(C=C)C=C1 |
Chemical Identifiers
| 1592-20-7 | |
| 152.62 | |
| ZRZHXNCATOYMJH-UHFFFAOYSA-N | |
| 74126 | |
| ClCC1=CC=C(C=C)C=C1 |
| C9H9Cl | |
| MFCD00051362 | |
| 4-vinylbenzyl chloride, 1-chloromethyl-4-vinylbenzene, 4-chloromethylstyrene, 4-vinylbenzylchloride, benzene, 1-chloromethyl-4-ethenyl, 1-chloromethyl-4-ethenylbenzene, 4-chloromethyl styrene, p-vinylbenzyl chloride, unii-cjl4oj6bs0, cjl4oj6bs0 | |
| 1-(chloromethyl)-4-ethenylbenzene |
Specifications
| 1592-20-7 | |
| 229°C | |
| MFCD00051362 | |
| 3265 | |
| ZRZHXNCATOYMJH-UHFFFAOYSA-N | |
| 1-(chloromethyl)-4-ethenylbenzene | |
| 74126 | |
| ≥90.0% (GC) | |
| 4-(Chloromethyl)styrene (stabilized with TBC + ONP + 2-Nitro-p-cresol) |
| Yellow | |
| C9H9Cl | |
| 25 g | |
| 4-vinylbenzyl chloride, 1-chloromethyl-4-vinylbenzene, 4-chloromethylstyrene, 4-vinylbenzylchloride, benzene, 1-chloromethyl-4-ethenyl, 1-chloromethyl-4-ethenylbenzene, 4-chloromethyl styrene, p-vinylbenzyl chloride, unii-cjl4oj6bs0, cjl4oj6bs0 | |
| ClCC1=CC=C(C=C)C=C1 | |
| 152.62 | |
| 152.62 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-0046
TSCA : Yes
Recommended Storage : Freezer