missing translation for 'onlineSavingsMsg'
Learn More

4-(Chloromethyl)styrene (stabilized with TBC + ONP + 2-Nitro-p-cresol) 90.0+%, TCI America™

Catalog No. C179225G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C179225G 25 g
C1792250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C179225G Supplier TCI America Supplier No. C179225G

Chemical Identifiers

CAS 1592-20-7
Molecular Formula C9H9Cl
Molecular Weight (g/mol) 152.62
MDL Number MFCD00051362
InChI Key ZRZHXNCATOYMJH-UHFFFAOYSA-N
Synonym 4-vinylbenzyl chloride, 1-chloromethyl-4-vinylbenzene, 4-chloromethylstyrene, 4-vinylbenzylchloride, benzene, 1-chloromethyl-4-ethenyl, 1-chloromethyl-4-ethenylbenzene, 4-chloromethyl styrene, p-vinylbenzyl chloride, unii-cjl4oj6bs0, cjl4oj6bs0
PubChem CID 74126
IUPAC Name 1-(chloromethyl)-4-ethenylbenzene
SMILES ClCC1=CC=C(C=C)C=C1

Specifications

Color Yellow
Boiling Point 229°C
Quantity 25 g
UN Number 3265
Formula Weight 152.62
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-(Chloromethyl)styrene (stabilized with TBC + ONP + 2-Nitro-p-cresol)
EINECSNumber (3)-0046
TSCA Yes
Recommended Storage Freezer