missing translation for 'onlineSavingsMsg'
Learn More

4-(Chloromethyl)phenyltrichlorosilane, 97%

Catalog No. p-7046812
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1643106 5 g
AAL1643114 25 g
AAL1643122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1643106 Supplier Thermo Scientific Chemicals Supplier No. L1643106
Only null left

CAS: 13688-90-9 | C7H6Cl4Si | 260.01 g/mol

4-(Chloromethyl)phenyltrichlorosilane is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Chloromethyl)phenyltrichlorosilane is used as a pharmaceutical intermediate.

Solubility
Reacts with water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas. Incompatible with oxidizing agents and bases. It is sensitive to moisture.

Chemical Identifiers

CAS 13688-90-9
Molecular Formula C7H6Cl4Si
Molecular Weight (g/mol) 260.01
MDL Number MFCD00045268
InChI Key YBYBQFPZMKPGPJ-UHFFFAOYSA-N
Synonym 4-chloromethyl phenyltrichlorosilane, p-chloromethyl phenyltrichlorosilane, silane, trichloro 4-chloromethyl phenyl, trichloro 4-chloromethyl phenyl silane, silane, trichloro .alpha.-chloro-p-tolyl, benzene, 1-chloromethyl-4-trichlorosilyl, p-chloromethylphenyltrichlorosilane
PubChem CID 83663
IUPAC Name trichloro-[4-(chloromethyl)phenyl]silane
SMILES C1=CC(=CC=C1CCl)[Si](Cl)(Cl)Cl

Specifications

Boiling Point 142°C to 144°C (15mmHg)
Quantity 5 g
UN Number UN2987
Beilstein 3090295
Flash Point 78°C (172°F)
Solubility Information Reacts with water.
Formula Weight 260.02
Percent Purity 97%
Refractive Index 1.548
Sensitivity Moisture sensitive; May form a precipitate
Density 1.361
Chemical Name or Material 4-(Chloromethyl)phenyltrichlorosilane
Show More Show Less
Hazard Category H227-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.
EINECSNumber 237-203-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only