CAS RN 13688-90-9
IUPAC Name:
trichloro[4-(chloromethyl)phenyl]silane
Synonyms:
trichloro[4-(chloromethyl)phenyl]silane
Molecular Weight (g/mol):
260.01
Molecular Formula:
C7H6Cl4Si
InChi Key:
YBYBQFPZMKPGPJ-UHFFFAOYSA-N
SMILES:
ClCC1=CC=C(C=C1)[Si](Cl)(Cl)Cl
1
–
1
of
1
results
1
4-(Chloromethyl)phenyltrichlorosilane, 97%
CAS: 13688-90-9 Molecular Formula: C7H6Cl4Si Molecular Weight (g/mol): 260.01 MDL Number: MFCD00045268 InChI Key: YBYBQFPZMKPGPJ-UHFFFAOYSA-N Synonym: 4-chloromethyl phenyltrichlorosilane,p-chloromethyl phenyltrichlorosilane,silane, trichloro 4-chloromethyl phenyl,trichloro 4-chloromethyl phenyl silane,silane, trichloro .alpha.-chloro-p-tolyl,benzene, 1-chloromethyl-4-trichlorosilyl,p-chloromethylphenyltrichlorosilane PubChem CID: 83663 IUPAC Name: trichloro-[4-(chloromethyl)phenyl]silane SMILES: C1=CC(=CC=C1CCl)[Si](Cl)(Cl)Cl