missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Chloromethyl)benzoyl Chloride 98.0+%, TCI America™
$123.21 - $571.77
Chemical Identifiers
| CAS | 876-08-4 |
|---|---|
| Molecular Formula | C8H6Cl2O |
| Molecular Weight (g/mol) | 189.035 |
| MDL Number | MFCD00053224 |
| InChI Key | RCOVTJVRTZGSBP-UHFFFAOYSA-N |
| Synonym | 4-chloromethyl benzoyl chloride, benzoyl chloride, 4-chloromethyl, 4-chloromethyl benzoylchloride, 4-chloromethyl-benzoyl chloride, acmc-209qop, 4-chlormethylbenzoylchloride, 4-chloromethylbenzoylchloride, 4-chlormethyl benzoylchlorid, p-chloromethylbenzoyl chloride |
| PubChem CID | 70136 |
| IUPAC Name | 4-(chloromethyl)benzoyl chloride |
| SMILES | C1=CC(=CC=C1CCl)C(=O)Cl |
Chemical Identifiers
| 876-08-4 | |
| 189.035 | |
| RCOVTJVRTZGSBP-UHFFFAOYSA-N | |
| 70136 | |
| C1=CC(=CC=C1CCl)C(=O)Cl |
| C8H6Cl2O | |
| MFCD00053224 | |
| 4-chloromethyl benzoyl chloride, benzoyl chloride, 4-chloromethyl, 4-chloromethyl benzoylchloride, 4-chloromethyl-benzoyl chloride, acmc-209qop, 4-chlormethylbenzoylchloride, 4-chloromethylbenzoylchloride, 4-chlormethyl benzoylchlorid, p-chloromethylbenzoyl chloride | |
| 4-(chloromethyl)benzoyl chloride |
Specifications
| 876-08-4 | |
| C8H6Cl2O | |
| 25 g | |
| 4-chloromethyl benzoyl chloride, benzoyl chloride, 4-chloromethyl, 4-chloromethyl benzoylchloride, 4-chloromethyl-benzoyl chloride, acmc-209qop, 4-chlormethylbenzoylchloride, 4-chloromethylbenzoylchloride, 4-chlormethyl benzoylchlorid, p-chloromethylbenzoyl chloride | |
| C1=CC(=CC=C1CCl)C(=O)Cl | |
| 189.035 | |
| 189.04 | |
| Crystalline Lumps |
| White | |
| MFCD00053224 | |
| 3261 | |
| RCOVTJVRTZGSBP-UHFFFAOYSA-N | |
| 4-(chloromethyl)benzoyl chloride | |
| 70136 | |
| ≥98.0% (GC,T) | |
| 4-(Chloromethyl)benzoyl Chloride |
Safety and Handling
EINECSNumber : (3)-4231
TSCA : No