missing translation for 'onlineSavingsMsg'
Learn More

4-(Chloromethyl)benzoyl Chloride 98.0+%, TCI America™

Catalog No. C128325G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C128325G 25 g
C1283100G 100 g
C1283500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C128325G Supplier TCI America Supplier No. C128325G

Chemical Identifiers

CAS 876-08-4
Molecular Formula C8H6Cl2O
Molecular Weight (g/mol) 189.035
MDL Number MFCD00053224
InChI Key RCOVTJVRTZGSBP-UHFFFAOYSA-N
Synonym 4-chloromethyl benzoyl chloride, benzoyl chloride, 4-chloromethyl, 4-chloromethyl benzoylchloride, 4-chloromethyl-benzoyl chloride, acmc-209qop, 4-chlormethylbenzoylchloride, 4-chloromethylbenzoylchloride, 4-chlormethyl benzoylchlorid, p-chloromethylbenzoyl chloride
PubChem CID 70136
IUPAC Name 4-(chloromethyl)benzoyl chloride
SMILES C1=CC(=CC=C1CCl)C(=O)Cl

Specifications

Color White
Quantity 25 g
UN Number 3261
Formula Weight 189.04
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 4-(Chloromethyl)benzoyl Chloride
EINECSNumber (3)-4231
TSCA No