missing translation for 'onlineSavingsMsg'
Learn More

4-(Chloromethyl)benzoic Acid 98.0+%, TCI America™

Catalog No. C121225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C121225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C121225G Supplier TCI America Supplier No. C121225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1642-81-5
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.592
MDL Number MFCD00002568
InChI Key OITNBJHJJGMFBN-UHFFFAOYSA-N
Synonym 4-chloromethyl benzoic acid, alpha-chloro-p-toluic acid, benzoic acid, 4-chloromethyl, 4-carboxybenzyl chloride, 4-chloromethyl-benzoic acid, alpha-chloro-p-toluylic acid, .alpha.-chloro-p-toluylic acid, pubchem15446, alpha-chloroparatoluic acid
PubChem CID 74234
IUPAC Name 4-(chloromethyl)benzoic acid
SMILES C1=CC(=CC=C1CCl)C(=O)O

Specifications

Melting Point 206°C
Color White
Quantity 25 g
UN Number 1759
Formula Weight 170.59
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Chloromethyl)benzoic Acid