missing translation for 'onlineSavingsMsg'
Learn More

4-(Chloromethyl)benzoic acid, 98%

Catalog No. AC392890500
Change view
Click to view available options
Quantity:
50 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC392890500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC392890500 Supplier Thermo Scientific Chemicals Supplier No. 392890500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 1642-81-5 | C8H7ClO2 | 170.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1642-81-5
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00002568
InChI Key OITNBJHJJGMFBN-UHFFFAOYSA-N
Synonym 4-chloromethyl benzoic acid, alpha-chloro-p-toluic acid, benzoic acid, 4-chloromethyl, 4-carboxybenzyl chloride, 4-chloromethyl-benzoic acid, alpha-chloro-p-toluylic acid, .alpha.-chloro-p-toluylic acid, pubchem15446, alpha-chloroparatoluic acid
PubChem CID 74234
IUPAC Name 4-(chloromethyl)benzoic acid
SMILES C1=CC(=CC=C1CCl)C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 202°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula ClCH2C6H4COOH
Quantity 50 g
Formula Weight 170.59
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-(Chloromethyl)benzoic acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 216-697-1

RUO – Research Use Only