missing translation for 'onlineSavingsMsg'
Learn More

4-Chloroindole 98.0+%, TCI America™

Catalog No. c12481g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C12481G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C12481G Supplier TCI America Supplier No. C12481G

Chemical Identifiers

CAS 25235-85-2
Molecular Formula C8H6ClN
Molecular Weight (g/mol) 151.59
MDL Number MFCD00005665
InChI Key SVLZRCRXNHITBY-UHFFFAOYSA-N
Synonym 4-chloroindole, 1h-indole, 4-chloro, 4-chloro indole, 1h-indole, 4-chloro-,conjugate acid 1:1, 4-chloro-indole, zlchem 277, indole, 4-chloro, pubchem7232, 4-chloranyl-1h-indole, acmc-209gia
PubChem CID 91345
IUPAC Name 4-chloro-1H-indole
SMILES ClC1=C2C=CNC2=CC=C1

Specifications

Color Yellow
Boiling Point 150°C
Quantity 1 g
Formula Weight 151.59
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Chloroindole