accessibility menu, dialog, popup

UserName

ClHN

CAS RN 25235-85-2

IUPAC Name: 4-chloro-1H-indole
Synonyms: 4-chloro-1H-indole
Molecular Weight (g/mol): 151.59
Molecular Formula: C8H6ClN
InChi Key: SVLZRCRXNHITBY-UHFFFAOYSA-N
SMILES: ClC1=C2C=CNC2=CC=C1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)