missing translation for 'onlineSavingsMsg'
Learn More

4-Chloroindole, 97%

Catalog No. p-7056923
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6615303 1 g
AAH6615306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6615303 Supplier Thermo Scientific Chemicals Supplier No. H6615303
Only null left

CAS: 25235-85-2 | C8H6ClN | 151.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25235-85-2
Molecular Formula C8H6ClN
Molecular Weight (g/mol) 151.59
MDL Number MFCD00005665
InChI Key SVLZRCRXNHITBY-UHFFFAOYSA-N
Synonym 4-chloroindole, 1h-indole, 4-chloro, 4-chloro indole, 1h-indole, 4-chloro-,conjugate acid 1:1, 4-chloro-indole, zlchem 277, indole, 4-chloro, pubchem7232, 4-chloranyl-1h-indole, acmc-209gia
PubChem CID 91345
IUPAC Name 4-chloro-1H-indole
SMILES ClC1=C2C=CNC2=CC=C1

Specifications

Density 1.259 g/mL
Boiling Point 129°C to 130°C (4 mmHg)
Flash Point 113°C (235°F)
Odor Odorless
Refractive Index 1.628
Quantity 1 g
Beilstein 114880
Formula Weight 151.59
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Chloroindole
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 246-747-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only