missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorodiphenylmethane 96.0+%, TCI America™

Catalog No. p-7125278
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20865G 5 g
C208625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C20865G Supplier TCI America Supplier No. C20865G
Only null left

Chemical Identifiers

CAS 831-81-2
Molecular Formula C13H11Cl
Molecular Weight (g/mol) 202.681
MDL Number MFCD00094070
InChI Key NPOGRKGIBGKRNI-UHFFFAOYSA-N
PubChem CID 13253
IUPAC Name 1-benzyl-4-chlorobenzene
SMILES C1=CC=C(C=C1)CC2=CC=C(C=C2)Cl

Specifications

Melting Point 7°C
Color Yellow
Boiling Point 153°C
Quantity 5 g
Formula Weight 202.68
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Chlorodiphenylmethane