missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorocinnamamide, 97%

Catalog No. AAL1140603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1140603 1 g
AAL1140606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1140603 Supplier Thermo Scientific Chemicals Supplier No. L1140603

CAS: 18166-64-8 | C9H8ClNO | 181.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18166-64-8
Molecular Formula C9H8ClNO
Molecular Weight (g/mol) 181.62
MDL Number MFCD00017147
InChI Key PWXPFYVNYKVJBW-ZZXKWVIFSA-N
Synonym 4-chlorocinnamamide, p-chlorocinnamamide, 2e-3-4-chlorophenyl prop-2-enamide, 3-4-chlorophenyl acrylamide, p-chlorocinnamide, 4-chlorocinnamide,trans, 4-chlorobenzeneacrylamide, 2e-3-4-chlorophenyl acrylamide, e-3-4-chlorophenyl prop-2-enamide
PubChem CID 5364144
IUPAC Name (E)-3-(4-chlorophenyl)prop-2-enamide
SMILES NC(=O)\C=C\C1=CC=C(Cl)C=C1

Specifications

Melting Point 210°C to 212°C
Quantity 1 g
Beilstein 2084774
Formula Weight 181.62
Percent Purity 97%
Chemical Name or Material 4-Chlorocinnamamide
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only