CAS RN 18166-64-8
IUPAC Name:
(2E)-3-(4-chlorophenyl)prop-2-enamide
Synonyms:
(2E)-3-(4-chlorophenyl)prop-2-enamide
Molecular Weight (g/mol):
181.62
Molecular Formula:
C9H8ClNO
InChi Key:
PWXPFYVNYKVJBW-ZZXKWVIFSA-N
SMILES:
NC(=O)\C=C\C1=CC=C(Cl)C=C1
1
–
1
of
1
results
1
Promotions Available
4-Chlorocinnamamide, 97%
4-Chlorocinnamamide, 97%
CAS: 18166-64-8 Molecular Formula: C9H8ClNO Molecular Weight (g/mol): 181.62 MDL Number: MFCD00017147 InChI Key: PWXPFYVNYKVJBW-ZZXKWVIFSA-N Synonym: 4-chlorocinnamamide,p-chlorocinnamamide,2e-3-4-chlorophenyl prop-2-enamide,3-4-chlorophenyl acrylamide,p-chlorocinnamide,4-chlorocinnamide,trans,4-chlorobenzeneacrylamide,2e-3-4-chlorophenyl acrylamide,e-3-4-chlorophenyl prop-2-enamide PubChem CID: 5364144 IUPAC Name: (E)-3-(4-chlorophenyl)prop-2-enamide SMILES: NC(=O)\C=C\C1=CC=C(Cl)C=C1