missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chlorocatechol 98.0+%, TCI America™
$357.90 - $1069.97
Chemical Identifiers
| CAS | 2138-22-9 |
|---|---|
| Molecular Formula | C6H5ClO2 |
| Molecular Weight (g/mol) | 144.554 |
| MDL Number | MFCD00059614 |
| InChI Key | WWOBYPKUYODHDG-UHFFFAOYSA-N |
| Synonym | 4-Chloro-1,2-dihydroxybenzene |
| PubChem CID | 16496 |
| ChEBI | CHEBI:27772 |
| IUPAC Name | 4-chlorobenzene-1,2-diol |
| SMILES | C1=CC(=C(C=C1Cl)O)O |
Chemical Identifiers
| 2138-22-9 | |
| 144.554 | |
| WWOBYPKUYODHDG-UHFFFAOYSA-N | |
| 16496 | |
| 4-chlorobenzene-1,2-diol |
| C6H5ClO2 | |
| MFCD00059614 | |
| 4-Chloro-1,2-dihydroxybenzene | |
| CHEBI:27772 | |
| C1=CC(=C(C=C1Cl)O)O |
Specifications
| 2138-22-9 | |
| 140°C | |
| MFCD00059614 | |
| 1759 | |
| WWOBYPKUYODHDG-UHFFFAOYSA-N | |
| 4-chlorobenzene-1,2-diol | |
| 16496 | |
| 144.55 | |
| Crystalline Powder |
| 88°C | |
| C6H5ClO2 | |
| 5 g | |
| 4-Chloro-1,2-dihydroxybenzene | |
| C1=CC(=C(C=C1Cl)O)O | |
| 144.554 | |
| CHEBI:27772 | |
| ≥98.0% (GC) | |
| 4-Chlorocatechol |
Safety and Handling
RTECSNumber : UX1490000
TSCA : No