missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobutyric Acid 95.0+%, TCI America™

Catalog No. p-7124545
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C065125G 25 g
C0651500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C065125G Supplier TCI America Supplier No. C065125G
Only null left

Chemical Identifiers

CAS 627-00-9
Molecular Formula C4H7ClO2
Molecular Weight (g/mol) 122.548
MDL Number MFCD00002818
InChI Key IPLKGJHGWCVSOG-UHFFFAOYSA-N
Synonym 4-chlorobutyric acid, butanoic acid, 4-chloro, butyric acid, 4-chloro, gamma-chlorobutyric acid, 4-chlorobutanoicacid, gammachlorobutyric acid, 4-chloro-butyric acid, 4-chloro-n-butyric acid, .gamma.-chlorobutyric acid, 4-chloranylbutanoic acid
PubChem CID 12300
IUPAC Name 4-chlorobutanoic acid
SMILES C(CC(=O)O)CCl

Specifications

Color Yellow
Boiling Point 125°C
Quantity 25 g
UN Number 3265
Formula Weight 122.55
Percent Purity ≥95.0% (T)
Physical Form Liquid
Chemical Name or Material 4-Chlorobutyric Acid