CAS RN 627-00-9
CAS RN 627-00-9
IUPAC Name:
4-chlorobutanoic acid
Synonyms:
butanoic acid, 4-chloro-
Molecular Weight (g/mol):
122.55
Molecular Formula:
C4H7ClO2
InChi Key:
IPLKGJHGWCVSOG-UHFFFAOYSA-N
SMILES:
OC(=O)CCCCl
1
–
1
of
1
results
1
4-Chlorobutyric Acid 95.0+%, TCI America™
CAS: 627-00-9 Molecular Formula: C4H7ClO2 Molecular Weight (g/mol): 122.548 MDL Number: MFCD00002818 InChI Key: IPLKGJHGWCVSOG-UHFFFAOYSA-N Synonym: 4-chlorobutyric acid,butanoic acid, 4-chloro,butyric acid, 4-chloro,gamma-chlorobutyric acid,4-chlorobutanoicacid,gammachlorobutyric acid,4-chloro-butyric acid,4-chloro-n-butyric acid,.gamma.-chlorobutyric acid,4-chloranylbutanoic acid PubChem CID: 12300 IUPAC Name: 4-chlorobutanoic acid SMILES: C(CC(=O)O)CCl