missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chlorobutyric Acid 95.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 125°C |
| Quantity | 500 g |
| UN Number | 3265 |
| Formula Weight | 122.55 |
| Percent Purity | ≥95.0% (T) |
| Physical Form | Liquid |
| Chemical Name or Material | 4-Chlorobutyric Acid |
Safety and Handling
| TSCA | Yes |