missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chlorobutyraldehyde Diethyl Acetal 97.0+%, TCI America™
$78.29 - $253.51
Chemical Identifiers
| CAS | 6139-83-9 |
|---|---|
| Molecular Formula | C8H17ClO2 |
| Molecular Weight (g/mol) | 180.672 |
| MDL Number | MFCD00671476 |
| InChI Key | JGGRHRMHOUWCDX-UHFFFAOYSA-N |
| Synonym | 4-Chloro-1,1-diethoxybutane |
| PubChem CID | 262598 |
| IUPAC Name | 4-chloro-1,1-diethoxybutane |
| SMILES | CCOC(CCCCl)OCC |
Chemical Identifiers
| 6139-83-9 | |
| 180.672 | |
| JGGRHRMHOUWCDX-UHFFFAOYSA-N | |
| 262598 | |
| CCOC(CCCCl)OCC |
| C8H17ClO2 | |
| MFCD00671476 | |
| 4-Chloro-1,1-diethoxybutane | |
| 4-chloro-1,1-diethoxybutane |
Specifications
| 6139-83-9 | |
| 92°C | |
| MFCD00671476 | |
| 1993 | |
| JGGRHRMHOUWCDX-UHFFFAOYSA-N | |
| 4-chloro-1,1-diethoxybutane | |
| 262598 | |
| ≥97.0% (GC) | |
| 4-Chlorobutyraldehyde Diethyl Acetal |
| Yellow | |
| C8H17ClO2 | |
| 5 g | |
| 4-Chloro-1,1-diethoxybutane | |
| CCOC(CCCCl)OCC | |
| 180.672 | |
| 180.67 | |
| Liquid |
Safety and Handling
TSCA : No