missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chlorobutyraldehyde Diethyl Acetal 97.0+%, TCI America™
Supplier: TCI America C271725G
Specifications
| 4-Chlorobutyraldehyde Diethyl Acetal | |
| Yellow | |
| C8H17ClO2 | |
| 25 g | |
| 4-Chloro-1,1-diethoxybutane | |
| CCOC(CCCCl)OCC | |
| 180.672 | |
| 180.67 | |
| Liquid |
| 6139-83-9 | |
| 92°C | |
| MFCD00671476 | |
| 1993 | |
| JGGRHRMHOUWCDX-UHFFFAOYSA-N | |
| 4-chloro-1,1-diethoxybutane | |
| 262598 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 6139-83-9 | |
| 180.672 | |
| JGGRHRMHOUWCDX-UHFFFAOYSA-N | |
| 262598 | |
| CCOC(CCCCl)OCC |
| C8H17ClO2 | |
| MFCD00671476 | |
| 4-Chloro-1,1-diethoxybutane | |
| 4-chloro-1,1-diethoxybutane |
Safety and Handling
TSCA : No