missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobutyl acetate, 98%

Catalog No. p-7026539
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2258214 25 g
AAB2258222 100 g
AAB2258236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2258214 Supplier Thermo Scientific Chemicals Supplier No. B2258214
Only null left

CAS: 6962-92-1 | C6H11ClO2 | 150.602 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6962-92-1
Molecular Formula C6H11ClO2
Molecular Weight (g/mol) 150.602
MDL Number MFCD00001013
InChI Key PYLDCZJUHYVOAF-UHFFFAOYSA-N
Synonym 1-acetoxy-4-chlorobutane, 4-chloro-n-butyl acetate, 1-chloro-4-acetoxybutane, 1-butanol, 4-chloro-, acetate, 4-chlorobutylacetate, 4-chloro-1-butanol acetate, 4-chlorbut-1-ylacetat, delta-chlorobutyl acetate, 1-butanol, 4-chloro-, 1-acetate, 4-chlorbut-1-ylacetat german
PubChem CID 23399
IUPAC Name 4-chlorobutyl acetate
SMILES CC(=O)OCCCCCl

Specifications

Density 1.076
Boiling Point 81°C to 83°C (11 mmHg)
Flash Point 64°C (147°F)
Refractive Index 1.435
Quantity 25 g
Beilstein 1749681
Formula Weight 150.61
Percent Purity 98%
Chemical Name or Material 4-Chlorobutyl acetate
Hazard Category H227-H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
EINECSNumber 230-158-8
RTECSNumber EL1328000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only