missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzylideneacetone, 98%

Catalog No. AAL0532114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0532114 25 g
AAL0532106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0532114 Supplier Thermo Scientific Chemicals Supplier No. L0532114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3160-40-5 | C10H9ClO | 180.631 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3160-40-5
Molecular Formula C10H9ClO
Molecular Weight (g/mol) 180.631
MDL Number MFCD00018790
InChI Key UUKRKWJGNHNTRG-NSCUHMNNSA-N
Synonym 4-chlorobenzylideneacetone, 4-chlorobenzalacetone, 4-4-chlorophenyl-3-buten-2-one, 4-4-chlorophenyl but-3-en-2-one, e-4-4-chlorophenyl but-3-en-2-one, 3e-4-4-chlorophenyl but-3-en-2-one, e-4-4-chlorophenyl-but-3-en-2-one, p-chlorobenzylidene acetone, 3-buten-2-one, 4-4-chlorophenyl, p-chlorobenzalacetone
PubChem CID 736572
IUPAC Name (E)-4-(4-chlorophenyl)but-3-en-2-one
SMILES CC(=O)C=CC1=CC=C(C=C1)Cl

Specifications

Melting Point 55°C to 60°C
Boiling Point 145°C to 147°C (5 mmHg)
Quantity 25 g
Beilstein 1100204
Formula Weight 180.64
Percent Purity 98%
Chemical Name or Material 4-Chlorobenzylideneacetone
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 221-610-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only