missing translation for 'onlineSavingsMsg'
Learn More

4-chlorobenzylamine, 98%

Catalog No. AC108790250
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC108790250 25 g Glass bottle
AC108790500 50 g Glass bottle
AC108791000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC108790250 Supplier Thermo Scientific Chemicals Supplier No. 108790250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 104-86-9 | C7H8ClN | 141.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 104-86-9
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.6
MDL Number MFCD00008121
InChI Key YMVFJGSXZNNUDW-UHFFFAOYSA-N
Synonym 4-chlorobenzylamine, 4-chlorophenyl methanamine, p-chlorobenzylamine, benzenemethanamine, 4-chloro, 1-4-chlorophenyl methanamine, 4-chloro-benzylamine, benzylamine, p-chloro, 4-chlorophenyl methylamine, para-chlorobenzylamine, chembl13218
PubChem CID 66036
IUPAC Name (4-chlorophenyl)methanamine
SMILES C1=CC(=CC=C1CN)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 1.1600g/mL
Boiling Point 215.0°C
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H4CH2NH2
Refractive Index 1.5576 to 1.5610
Quantity 25 g
Beilstein 12, 1074
Specific Gravity 1.16
Solubility Information Solubility in water: immiscible with water
Formula Weight 141.6
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-chlorobenzylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-245-3

RUO – Research Use Only