missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chlorobenzylamine, 97+%
Description
4-Chlorobenzylamine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Chlorobenzylamine is used as pharmaceutical intermediate.
Solubility
Not miscible in water.
Notes
Air sensitive. Incompatible with oxidizing agents and carbon dioxide.
4-Chlorobenzylamine is used as pharmaceutical intermediate.
Solubility
Not miscible in water.
Notes
Air sensitive. Incompatible with oxidizing agents and carbon dioxide.
Specifications
Specifications
| Density | 1.16 |
| Boiling Point | 229°C |
| Flash Point | 90°C (194°F) |
| Odor | Amine-like |
| Refractive Index | 1.559 |
| Quantity | 100 g |
| UN Number | UN2735 |
| Beilstein | 507949 |
| Sensitivity | Air Sensitive |
| Solubility Information | Not miscible in water. |
| Show More |
Safety and Handling
| Hazard Category | H227-H302+H312+H332-H314-H335 |
| Hazard Statement | GHS H Statement H314-H318-H227 Causes severe skin burns and eye damage. Causes serious eye damage. Combustible liquid. |
| Precautionary Statement | P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S. |
| EINECSNumber | 203-245-3 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only