missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzylamine, 97+%

Catalog No. aaa1398414
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1398414 25 g
AAA1398406 5 g
AAA1398422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1398414 Supplier Thermo Scientific Chemicals Supplier No. A1398414
Only null left

CAS: 104-86-9 | C7H8ClN | 141.598 g/mol

4-Chlorobenzylamine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorobenzylamine is used as pharmaceutical intermediate.

Solubility
Not miscible in water.

Notes
Air sensitive. Incompatible with oxidizing agents and carbon dioxide.

Chemical Identifiers

CAS 104-86-9
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.598
MDL Number MFCD00008121
InChI Key YMVFJGSXZNNUDW-UHFFFAOYSA-N
Synonym 4-chlorobenzylamine, 4-chlorophenyl methanamine, p-chlorobenzylamine, benzenemethanamine, 4-chloro, 1-4-chlorophenyl methanamine, 4-chloro-benzylamine, benzylamine, p-chloro, 4-chlorophenyl methylamine, para-chlorobenzylamine, chembl13218
PubChem CID 66036
IUPAC Name (4-chlorophenyl)methanamine
SMILES C1=CC(=CC=C1CN)Cl

Specifications

Density 1.16
Boiling Point 229°C
Flash Point 90°C (194°F)
Odor Amine-like
Refractive Index 1.559
Quantity 25 g
UN Number UN2735
Beilstein 507949
Sensitivity Air Sensitive
Solubility Information Not miscible in water.
Formula Weight 141.6
Percent Purity ≥97%
Chemical Name or Material 4-Chlorobenzylamine
Show More Show Less
Hazard Category H227-H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 203-245-3
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only