missing translation for 'onlineSavingsMsg'
Learn More

(4-Chlorobenzyl)triphenylphosphonium chloride, 98+%

Catalog No. p-7081691
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1176914 25 g
AAA1176922 100 g
AAA1176936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1176914 Supplier Thermo Scientific Chemicals Supplier No. A1176914
Only null left

CAS: 1530-39-8 | C25H21Cl2P | 423.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1530-39-8
Molecular Formula C25H21Cl2P
Molecular Weight (g/mol) 423.32
MDL Number MFCD00041533
InChI Key RAHOAHBOOHXRDY-UHFFFAOYSA-M
Synonym 4-chlorobenzyl triphenylphosphonium chloride, 4-chlorobenzyl triphenylphosphoniumchloride, 4-chlorophenyl methyl triphenylphosphanium chloride, acmc-1bpq8, 4-chlorobenzyltriphenylphosphonium chloride, 4-chlorobenzyltriphenyl-phosphonium chloride, 4-chlorophenyl methyl-triphenylphosphanium chloride, 4-chlorophenyl methyl-triphenylphosphonium chloride, 4-chlorophenyl methyl triphenylphosphine, chloride, 4-chlorobenzyl triphenyl phosphonium chloride
PubChem CID 2733546
SMILES [Cl-].ClC1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1

Specifications

Melting Point 286°C to 291°C
Quantity 25 g
Beilstein 4173400
Sensitivity Hygroscopic
IUPAC Name (4-chlorophenyl)methyl-triphenylphosphanium;chloride
Formula Weight 423.32
Percent Purity ≥98%
Chemical Name or Material (4-Chlorobenzyl)triphenylphosphonium chloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 216-228-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only