missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzyl Chloride 98.0+%, TCI America™

Catalog No. D042125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D042125G 25 g
D0421500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D042125G Supplier TCI America Supplier No. D042125G

Chemical Identifiers

CAS 104-83-6
Molecular Formula C7H6Cl2
Molecular Weight (g/mol) 161.025
MDL Number MFCD00000914
InChI Key JQZAEUFPPSRDOP-UHFFFAOYSA-N
Synonym 4-chlorobenzyl chloride, 1-chloro-4-chloromethyl benzene, p-chlorobenzyl chloride, benzene, 1-chloro-4-chloromethyl, alpha,4-dichlorotoluene, 4-chlorobenzylchloride, 4-chlorophenyl methyl chloride, 1-chloro-4-chloromethyl-benzene, alpha,p-dichlorotoluene, toluene, p,alpha-dichloro
PubChem CID 7723
IUPAC Name 1-chloro-4-(chloromethyl)benzene
SMILES C1=CC(=CC=C1CCl)Cl

Specifications

Color White
Boiling Point 222°C
Quantity 25 g
UN Number 3427
Formula Weight 161.03
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Chlorobenzyl Chloride
EINECSNumber (3)-0078&(3)-0091
RTECSNumber XT0720000
TSCA Yes