missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chlorobenzyl chloride, 99+%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 99.0 |
| CAS Max % | 100.0 |
| Melting Point | 27°C to 28°C |
| Color | Colorless to White |
| Boiling Point | 221°C |
| Flash Point | 107°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 0.5% max. p-Chlorobenzal chloride (GC) |
| Packaging | Glass bottle |
| Linear Formula | ClC6H4CH2Cl |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Harmful in contact with skin. Harmful if inhaled. Toxic to aquatic life with long lasting effects. Harmful if swallowed. May cause an allergic skin reaction. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for severa |
| EINECSNumber | 203-242-7 |
| RTECSNumber | XT0720000 |
| TSCA | TSCA |
RUO – Research Use Only