missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chlorobenzyl Bromide 98.0+%, TCI America™
$39.14 - $167.76
Chemical Identifiers
| CAS | 622-95-7 |
|---|---|
| Molecular Formula | C7H6BrCl |
| Molecular Weight (g/mol) | 205.48 |
| MDL Number | MFCD00040714 |
| InChI Key | KQNBRMUBPRGXSL-UHFFFAOYSA-N |
| Synonym | 4-chlorobenzyl bromide, 1-bromomethyl-4-chlorobenzene, p-chlorobenzyl bromide, 4-chlorobenzylbromide, benzene, 1-bromomethyl-4-chloro, alpha-bromo-4-chlorotoluene, 4cbb, p-chlorobenzylbromide, 4-chloro-benzylbromide |
| PubChem CID | 69329 |
| IUPAC Name | 1-(bromomethyl)-4-chlorobenzene |
| SMILES | ClC1=CC=C(CBr)C=C1 |
Chemical Identifiers
| 622-95-7 | |
| 205.48 | |
| KQNBRMUBPRGXSL-UHFFFAOYSA-N | |
| 69329 | |
| ClC1=CC=C(CBr)C=C1 |
| C7H6BrCl | |
| MFCD00040714 | |
| 4-chlorobenzyl bromide, 1-bromomethyl-4-chlorobenzene, p-chlorobenzyl bromide, 4-chlorobenzylbromide, benzene, 1-bromomethyl-4-chloro, alpha-bromo-4-chlorotoluene, 4cbb, p-chlorobenzylbromide, 4-chloro-benzylbromide | |
| 1-(bromomethyl)-4-chlorobenzene |
Specifications
| 622-95-7 | |
| White-Yellow | |
| C7H6BrCl | |
| 5 g | |
| 4-chlorobenzyl bromide, 1-bromomethyl-4-chlorobenzene, p-chlorobenzyl bromide, 4-chlorobenzylbromide, benzene, 1-bromomethyl-4-chloro, alpha-bromo-4-chlorotoluene, 4cbb, p-chlorobenzylbromide, 4-chloro-benzylbromide | |
| ClC1=CC=C(CBr)C=C1 | |
| 205.48 | |
| 205.48 | |
| Crystalline Powder |
| 50°C | |
| 110°C | |
| MFCD00040714 | |
| 3261 | |
| KQNBRMUBPRGXSL-UHFFFAOYSA-N | |
| 1-(bromomethyl)-4-chlorobenzene | |
| 69329 | |
| ≥98.0% (GC) | |
| 4-Chlorobenzyl Bromide |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator