missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzyl bromide, 98%

Catalog No. AC391710050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC391710050 5 g Glass bottle
AC391710250 25 g Glass bottle
AC391711000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC391710050 Supplier Thermo Scientific Chemicals Supplier No. 391710050

CAS: 622-95-7 | C7H6BrCl | 205.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 622-95-7
Molecular Formula C7H6BrCl
Molecular Weight (g/mol) 205.48
MDL Number MFCD00040714
InChI Key KQNBRMUBPRGXSL-UHFFFAOYSA-N
Synonym 4-chlorobenzyl bromide, 1-bromomethyl-4-chlorobenzene, p-chlorobenzyl bromide, 4-chlorobenzylbromide, benzene, 1-bromomethyl-4-chloro, alpha-bromo-4-chlorotoluene, 4cbb, p-chlorobenzylbromide, 4-chloro-benzylbromide
PubChem CID 69329
IUPAC Name 1-(bromomethyl)-4-chlorobenzene
SMILES ClC1=CC=C(CBr)C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 46°C to 50°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrCH2C6H4Cl
Quantity 5 g
Formula Weight 205.48
Percent Purity 98%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 4-Chlorobenzyl bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-760-7

RUO – Research Use Only