missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzyl bromide, 98+%

CAS: 622-95-7 | C7H6BrCl | 205.48 g/mol

$95.85 - $259.49

Chemical Identifiers

CAS 622-95-7
Molecular Formula C7H6BrCl
Molecular Weight (g/mol) 205.48
MDL Number MFCD00040714
InChI Key KQNBRMUBPRGXSL-UHFFFAOYSA-N
Synonym 4-chlorobenzyl bromide, 1-bromomethyl-4-chlorobenzene, p-chlorobenzyl bromide, 4-chlorobenzylbromide, benzene, 1-bromomethyl-4-chloro, alpha-bromo-4-chlorotoluene, 4cbb, p-chlorobenzylbromide, 4-chloro-benzylbromide
PubChem CID 69329
IUPAC Name 1-(bromomethyl)-4-chlorobenzene
SMILES ClC1=CC=C(CBr)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2285806
View Documents
Thermo Scientific Chemicals
B2285806
5 g
Each for $95.85
Only null left
 
AAB2285814
View Documents
Thermo Scientific Chemicals
B2285814
25 g
Each for $259.49
Only null left
 
Description

Description

4-Chlorobenzyl bromide is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorobenzyl bromide is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, bases and active metals.
Specifications

Chemical Identifiers

622-95-7
205.48
KQNBRMUBPRGXSL-UHFFFAOYSA-N
69329
ClC1=CC=C(CBr)C=C1
C7H6BrCl
MFCD00040714
4-chlorobenzyl bromide, 1-bromomethyl-4-chlorobenzene, p-chlorobenzyl bromide, 4-chlorobenzylbromide, benzene, 1-bromomethyl-4-chloro, alpha-bromo-4-chlorotoluene, 4cbb, p-chlorobenzylbromide, 4-chloro-benzylbromide
1-(bromomethyl)-4-chlorobenzene

Specifications

622-95-7
107°C to 108°C (8 mmHg)
C7H6BrCl
5 g
606497
Insoluble in water.
ClC1=CC=C(CBr)C=C1
205.48
205.49
4-Chlorobenzyl bromide
46°C to 51°C
Pungent
MFCD00040714
UN3261
4-chlorobenzyl bromide, 1-bromomethyl-4-chlorobenzene, p-chlorobenzyl bromide, 4-chlorobenzylbromide, benzene, 1-bromomethyl-4-chloro, alpha-bromo-4-chlorotoluene, 4cbb, p-chlorobenzylbromide, 4-chloro-benzylbromide
KQNBRMUBPRGXSL-UHFFFAOYSA-N
1-(bromomethyl)-4-chlorobenzene
69329
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 210-760-7

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only