missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chlorobenzyl alcohol, 99%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Applications
4-Chlorobenzyl alcohol acts as a reagent for the protection of carboxyl groups as their 4-chlorobenzyl esters, more stable to acid than the corresponding benzyl esters. It is used as solvent in paint stripper and waterborne coatings. It acts as curing agent. It is also used in pharmaceuticals, cosmetics, preservatives, and flavoring & fragrance agents.
Solubility
Soluble in water (2.5 mg/ml at 20°C), and methanol.
Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
4-Chlorobenzyl alcohol acts as a reagent for the protection of carboxyl groups as their 4-chlorobenzyl esters, more stable to acid than the corresponding benzyl esters. It is used as solvent in paint stripper and waterborne coatings. It acts as curing agent. It is also used in pharmaceuticals, cosmetics, preservatives, and flavoring & fragrance agents.
Solubility
Soluble in water (2.5 mg/ml at 20°C), and methanol.
Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
Specifications
Specifications
| Melting Point | 68°C to 72°C |
| Boiling Point | 234°C |
| Quantity | 25 g |
| Beilstein | 636502 |
| Solubility Information | Soluble in water (2.5mg/ml at 20°C),and methanol. |
| Formula Weight | 142.59 |
| Percent Purity | 99% |
| Chemical Name or Material | 4-Chlorobenzyl alcohol |
Safety and Handling
| EINECSNumber | 212-852-2 |
| RTECSNumber | DN9242000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only