missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzyl alcohol, 99%

Catalog No. AAA1150422
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1150422 100 g
AAA1150414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1150422 Supplier Thermo Scientific Chemicals Supplier No. A1150422
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 873-76-7 | C7H7ClO | 142.582 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
4-Chlorobenzyl alcohol acts as a reagent for the protection of carboxyl groups as their 4-chlorobenzyl esters, more stable to acid than the corresponding benzyl esters. It is used as solvent in paint stripper and waterborne coatings. It acts as curing agent. It is also used in pharmaceuticals, cosmetics, preservatives, and flavoring & fragrance agents.

Solubility
Soluble in water (2.5 mg/ml at 20°C), and methanol.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.

Chemical Identifiers

CAS 873-76-7
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.582
MDL Number MFCD00004652
InChI Key PTHGDVCPCZKZKR-UHFFFAOYSA-N
Synonym 4-chlorobenzyl alcohol, 4-chlorophenyl methanol, p-chlorobenzyl alcohol, benzenemethanol, 4-chloro, 4-chlorobenzenemethanol, 4-chlorobenzylalcohol, benzyl alcohol, p-chloro, 4-chlorobenzylic alcohol, unii-gf8bs53b6v, ccris 5120
PubChem CID 13397
IUPAC Name (4-chlorophenyl)methanol
SMILES C1=CC(=CC=C1CO)Cl

Specifications

Melting Point 68°C to 72°C
Boiling Point 234°C
Quantity 100 g
Beilstein 636502
Solubility Information Soluble in water (2.5mg/ml at 20°C),and methanol.
Formula Weight 142.59
Percent Purity 99%
Chemical Name or Material 4-Chlorobenzyl alcohol
EINECSNumber 212-852-2
RTECSNumber DN9242000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only