missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chlorobenzoyl-L-tryptophan Calcium Salt 98.0+%, TCI America™
$283.33 - $846.29
Chemical Identifiers
| CAS | 56116-62-2 |
|---|---|
| Molecular Formula | C36H28Ca2Cl2N4O6+2 |
| Molecular Weight (g/mol) | 763.698 |
| MDL Number | MFCD00036700 |
| InChI Key | RTNAGHDBACTRBP-OLYXIJPGSA-L |
| PubChem CID | 133126624 |
| IUPAC Name | dicalcium;(2R)-2-[(4-chlorobenzoyl)amino]-3-(1H-indol-3-yl)propanoate |
| SMILES | C1=CC=C2C(=C1)C(=CN2)CC(C(=O)[O-])NC(=O)C3=CC=C(C=C3)Cl.C1=CC=C2C(=C1)C(=CN2)CC(C(=O)[O-])NC(=O)C3=CC=C(C=C3)Cl.[Ca+2].[Ca+2] |
Chemical Identifiers
| 56116-62-2 | |
| 763.698 | |
| RTNAGHDBACTRBP-OLYXIJPGSA-L | |
| dicalcium;(2R)-2-[(4-chlorobenzoyl)amino]-3-(1H-indol-3-yl)propanoate |
| C36H28Ca2Cl2N4O6+2 | |
| MFCD00036700 | |
| 133126624 | |
| C1=CC=C2C(=C1)C(=CN2)CC(C(=O)[O-])NC(=O)C3=CC=C(C=C3)Cl.C1=CC=C2C(=C1)C(=CN2)CC(C(=O)[O-])NC(=O)C3=CC=C(C=C3)Cl.[Ca+2].[Ca+2] |
Specifications
| 56116-62-2 | |
| C36H28Ca2Cl2N4O6+2 | |
| 5 g | |
| C1=CC=C2C(=C1)C(=CN2)CC(C(=O)[O-])NC(=O)C3=CC=C(C=C3)Cl.C1=CC=C2C(=C1)C(=CN2)CC(C(=O)[O-])NC(=O)C3=CC=C(C=C3)Cl.[Ca+2].[Ca+2] | |
| 763.698 | |
| 723.62 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD00036700 | |
| RTNAGHDBACTRBP-OLYXIJPGSA-L | |
| dicalcium;(2R)-2-[(4-chlorobenzoyl)amino]-3-(1H-indol-3-yl)propanoate | |
| 133126624 | |
| ≥98.0% (HPLC) | |
| 4-Chlorobenzoyl-L-tryptophan Calcium Salt |
Safety and Handling
TSCA : No