accessibility menu, dialog, popup

UserName

CAS RN 56116-62-2

CaOO(S)HNNHOClOO(S)NHNHOCl

CAS RN 56116-62-2

IUPAC Name: calcium bis((2S)-2-[(4-chlorophenyl)formamido]-3-(1H-indol-3-yl)propanoate)
Synonyms: calcium bis((2S)-2-[(4-chlorophenyl)formamido]-3-(1H-indol-3-yl)propanoate)
Molecular Weight (g/mol): 723.62
Molecular Formula: C36H28CaCl2N4O6
InChi Key: BTTZKEFKFLGNEC-WUBQCMAVSA-L
SMILES: [Ca++].[O-]C(=O)[C@H](CC1=CNC2=CC=CC=C12)NC(=O)C1=CC=C(Cl)C=C1.[O-]C(=O)[C@H](CC1=CNC2=CC=CC=C12)NC(=O)C1=CC=C(Cl)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results