missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzoyl Chloride 98.0+%, TCI America™

Supplier:  TCI America C0141500G

 View more versions of this product

Catalog No. C0141500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4-Chlorobenzoyl Chloride
Colorless
C7H4Cl2O
500 g
benzoyl chloride, 4-chloro, p-chlorobenzoyl chloride, para-chlorobenzoyl chloride, 4-chlorobenzoic acid chloride, 4-chlorobenzoylchloride, 4-chloro benzoyl chloride, benzoylchloride, 4-chloro, benzoyl chloride, p-chloro, ccris 8935, pcboc
C1=CC(=CC=C1C(=O)Cl)Cl
175.008
CHEBI:60716
≥98.0% (GC,T)
122-01-0
222°C
MFCD00000686
3265
RKIDDEGICSMIJA-UHFFFAOYSA-N
4-chlorobenzoyl chloride
8501
175.01
Liquid

Chemical Identifiers

122-01-0
175.008
RKIDDEGICSMIJA-UHFFFAOYSA-N
8501
4-chlorobenzoyl chloride
C7H4Cl2O
MFCD00000686
benzoyl chloride, 4-chloro, p-chlorobenzoyl chloride, para-chlorobenzoyl chloride, 4-chlorobenzoic acid chloride, 4-chlorobenzoylchloride, 4-chloro benzoyl chloride, benzoylchloride, 4-chloro, benzoyl chloride, p-chloro, ccris 8935, pcboc
CHEBI:60716
C1=CC(=CC=C1C(=O)Cl)Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0062&(3)-1409

RTECSNumber : DM6635510

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions