missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzoyl chloride, 99+%

Catalog No. p-4510155
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC150191000 100 mL Glass bottle
AC150195000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC150191000 Supplier Thermo Scientific Chemicals Supplier No. 150191000

CAS: 122-01-0 | C7H4Cl2O | 175.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 122-01-0
Molecular Formula C7H4Cl2O
Molecular Weight (g/mol) 175.01
InChI Key RKIDDEGICSMIJA-UHFFFAOYSA-N
Synonym benzoyl chloride, 4-chloro, p-chlorobenzoyl chloride, para-chlorobenzoyl chloride, 4-chlorobenzoic acid chloride, 4-chlorobenzoylchloride, 4-chloro benzoyl chloride, benzoylchloride, 4-chloro, benzoyl chloride, p-chloro, ccris 8935, pcboc
PubChem CID 8501
ChEBI CHEBI:60716
IUPAC Name 4-chlorobenzoyl chloride
SMILES C1=CC(=CC=C1C(=O)Cl)Cl

Specifications

Melting Point 12.0°C to 14.0°C
Color Colorless
Density 1.3700g/mL
Boiling Point 222.0°C
Flash Point 105°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.5770 to 1.5790
Linear Formula ClC6H4COCl
Quantity 100 mL
Beilstein 09,341
Specific Gravity 1.37
Solubility Information Solubility in water: hydrolyses. Other solubilities: soluble in alcohol,ether and acetone
Formula Weight 175.01
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 4-Chlorobenzoyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 204-515-3

RUO – Research Use Only