missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzophenone 98.0+%, TCI America™

Catalog No. c013625g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
C013625G 25 g
C0136500G 500 g
2 options

Catalog No. C013625G

Supplier: TCI America C013625G

Only null left

Chemical Identifiers

CAS 134-85-0
Molecular Formula C13H9ClO
Molecular Weight (g/mol) 216.664
MDL Number MFCD00000622
InChI Key UGVRJVHOJNYEHR-UHFFFAOYSA-N
Synonym 4-chlorobenzophenone, 4-chlorophenyl phenyl methanone, p-chlorobenzophenone, methanone, 4-chlorophenyl phenyl, benzophenone, 4-chloro, p-cbp, unii-wih1iz728u, 4-chloro benzophenone, 4-chlorophenyl-phenylmethanone, wih1iz728u
PubChem CID 8653
IUPAC Name (4-chlorophenyl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)Cl

Specifications

Melting Point 76°C
Color White
Boiling Point 170°C
Quantity 25 g
Formula Weight 216.66
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chlorobenzophenone
EINECSNumber (4)-0127
TSCA Yes