missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzophenone, 99%

Catalog No. p-7023663
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1745822 100 g
AAA1745836 500 g
AAA174580E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1745822 Supplier Thermo Scientific Chemicals Supplier No. A1745822

CAS: 134-85-0 | C13H9ClO | 216.664 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 134-85-0
Molecular Formula C13H9ClO
Molecular Weight (g/mol) 216.664
MDL Number MFCD00000622
InChI Key UGVRJVHOJNYEHR-UHFFFAOYSA-N
Synonym 4-chlorobenzophenone, 4-chlorophenyl phenyl methanone, p-chlorobenzophenone, methanone, 4-chlorophenyl phenyl, benzophenone, 4-chloro, p-cbp, unii-wih1iz728u, 4-chloro benzophenone, 4-chlorophenyl-phenylmethanone, wih1iz728u
PubChem CID 8653
IUPAC Name (4-chlorophenyl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)Cl

Specifications

Melting Point 75°C to 77°C
Boiling Point 332°C
Flash Point 143°C (289°F)
Quantity 100 g
Beilstein 512043
Formula Weight 216.67
Percent Purity 99%
Chemical Name or Material 4-Chlorobenzophenone
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 205-160-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only