missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzonitrile 98.0+%, TCI America™

Supplier:  TCI America C0135500G

 View more versions of this product

Catalog No. C0135500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4-Chlorobenzonitrile
623-03-0
223°C
Crystal-Powder at 20°C
MFCD00001813
GJNGXPDXRVXSEH-UHFFFAOYSA-N
4-chlorobenzonitrile
12163
≥98.0% (GC)
92°C
White-Yellow
500 g
C7H4ClN
p-chlorobenzonitrile, benzonitrile, 4-chloro, p-cyanochlorobenzene, benzonitrile, p-chloro, 4-cyanochlorobenzene, nitril kyseliny p-chlorbenzoove, p-chlorbenzonitrile, unii-4z0hgp3a8a, pcbn, 4-chloro benzonitrile
ClC1=CC=C(C=C1)C#N
137.57
137.57

Chemical Identifiers

623-03-0
137.57
GJNGXPDXRVXSEH-UHFFFAOYSA-N
12163
ClC1=CC=C(C=C1)C#N
C7H4ClN
MFCD00001813
p-chlorobenzonitrile, benzonitrile, 4-chloro, p-cyanochlorobenzene, benzonitrile, p-chloro, 4-cyanochlorobenzene, nitril kyseliny p-chlorbenzoove, p-chlorbenzonitrile, unii-4z0hgp3a8a, pcbn, 4-chloro benzonitrile
4-chlorobenzonitrile
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1785

RTECSNumber : DI2800000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions