missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzonitrile 98.0+%, TCI America™

Catalog No. p-7124237
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C013525G 25 g
C0135500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C013525G Supplier TCI America Supplier No. C013525G
Only null left

Chemical Identifiers

CAS 623-03-0
Molecular Formula C7H4ClN
Molecular Weight (g/mol) 137.57
MDL Number MFCD00001813
InChI Key GJNGXPDXRVXSEH-UHFFFAOYSA-N
Synonym p-chlorobenzonitrile, benzonitrile, 4-chloro, p-cyanochlorobenzene, benzonitrile, p-chloro, 4-cyanochlorobenzene, nitril kyseliny p-chlorbenzoove, p-chlorbenzonitrile, unii-4z0hgp3a8a, pcbn, 4-chloro benzonitrile
PubChem CID 12163
IUPAC Name 4-chlorobenzonitrile
SMILES ClC1=CC=C(C=C1)C#N

Specifications

Melting Point 92°C
Color White-Yellow
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Boiling Point 223°C
Formula Weight 137.57
Percent Purity ≥98.0% (GC)
Chemical Name or Material 4-Chlorobenzonitrile
EINECSNumber (3)-1785
RTECSNumber DI2800000
TSCA Yes