missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzonitrile, 99%

CAS: 623-03-0 | C7H4ClN | 137.57 g/mol

$136.97 - $1,585.25

Chemical Identifiers

CAS 623-03-0
Molecular Formula C7H4ClN
Molecular Weight (g/mol) 137.57
MDL Number MFCD00001813
InChI Key GJNGXPDXRVXSEH-UHFFFAOYSA-N
Synonym p-chlorobenzonitrile, benzonitrile, 4-chloro, p-cyanochlorobenzene, benzonitrile, p-chloro, 4-cyanochlorobenzene, nitril kyseliny p-chlorbenzoove, p-chlorbenzonitrile, unii-4z0hgp3a8a, pcbn, 4-chloro benzonitrile
PubChem CID 12163
IUPAC Name 4-chlorobenzonitrile
SMILES ClC1=CC=C(C=C1)C#N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1452418
View Documents
Thermo Scientific Chemicals
A1452418
50 g N/A
Only null left
 
AAA1452430
View Documents
Thermo Scientific Chemicals
A1452430
250 g N/A
Only null left
 
AAA145240B
View Documents
Thermo Scientific Chemicals
A145240B
1000 g N/A
Only null left
 
Description

Description

4-Chlorobenzonitrile is used to prepare a red pigment for plastics. It is also used in the preparation of amides in organic synthesis. Further, it is used as an intermediate of dye and active pharmaceutical ingredients.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorobenzonitrile is used to prepare a red pigment for plastics. It is also used in the preparation of amides in organic synthesis. Further, it is used as an intermediate of dye and active pharmaceutical ingredients.

Solubility
Soluble in alcohol and ether. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

623-03-0
137.57
GJNGXPDXRVXSEH-UHFFFAOYSA-N
12163
ClC1=CC=C(C=C1)C#N
C7H4ClN
MFCD00001813
p-chlorobenzonitrile, benzonitrile, 4-chloro, p-cyanochlorobenzene, benzonitrile, p-chloro, 4-cyanochlorobenzene, nitril kyseliny p-chlorbenzoove, p-chlorbenzonitrile, unii-4z0hgp3a8a, pcbn, 4-chloro benzonitrile
4-chlorobenzonitrile

Specifications

90°C to 94°C
1.2
50 g
C7H4ClN
1072122
Soluble in alcohol and ether. Insoluble in water.
ClC1=CC=C(C=C1)C#N
137.57
137.57
4-Chlorobenzonitrile
623-03-0
222°C to 224°C
Almond-like
MFCD00001813
p-chlorobenzonitrile, benzonitrile, 4-chloro, p-cyanochlorobenzene, benzonitrile, p-chloro, 4-cyanochlorobenzene, nitril kyseliny p-chlorbenzoove, p-chlorbenzonitrile, unii-4z0hgp3a8a, pcbn, 4-chloro benzonitrile
GJNGXPDXRVXSEH-UHFFFAOYSA-N
4-chlorobenzonitrile
12163
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H319
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.

P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P312-P330-P363-P501c

H302+H312-H319

EINECSNumber : 210-765-4

RTECSNumber : DI2800000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only