missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzonitrile, 99%

CAS: 623-03-0 | C7H4ClN | 137.57 g/mol

Supplier:  Thermo Scientific Chemicals A145240B

 View more versions of this product

Catalog No. AAA145240B

Only null left
Explore available promotions

Description

Description

4-Chlorobenzonitrile is used to prepare a red pigment for plastics. It is also used in the preparation of amides in organic synthesis. Further, it is used as an intermediate of dye and active pharmaceutical ingredients.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorobenzonitrile is used to prepare a red pigment for plastics. It is also used in the preparation of amides in organic synthesis. Further, it is used as an intermediate of dye and active pharmaceutical ingredients.

Solubility
Soluble in alcohol and ether. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

4-Chlorobenzonitrile
623-03-0
222°C to 224°C
Almond-like
MFCD00001813
p-chlorobenzonitrile, benzonitrile, 4-chloro, p-cyanochlorobenzene, benzonitrile, p-chloro, 4-cyanochlorobenzene, nitril kyseliny p-chlorbenzoove, p-chlorbenzonitrile, unii-4z0hgp3a8a, pcbn, 4-chloro benzonitrile
GJNGXPDXRVXSEH-UHFFFAOYSA-N
4-chlorobenzonitrile
12163
99%
90°C to 94°C
1.2
1000 g
C7H4ClN
1072122
Soluble in alcohol and ether. Insoluble in water.
ClC1=CC=C(C=C1)C#N
137.57
137.57

Chemical Identifiers

623-03-0
137.57
GJNGXPDXRVXSEH-UHFFFAOYSA-N
12163
ClC1=CC=C(C=C1)C#N
C7H4ClN
MFCD00001813
p-chlorobenzonitrile, benzonitrile, 4-chloro, p-cyanochlorobenzene, benzonitrile, p-chloro, 4-cyanochlorobenzene, nitril kyseliny p-chlorbenzoove, p-chlorbenzonitrile, unii-4z0hgp3a8a, pcbn, 4-chloro benzonitrile
4-chlorobenzonitrile
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H319
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.

P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P312-P330-P363-P501c

H302+H312-H319

EINECSNumber : 210-765-4

RTECSNumber : DI2800000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only