missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chlorobenzohydrazide 98.0+%, TCI America™
$93.21 - $372.81
Chemical Identifiers
| CAS | 536-40-3 |
|---|---|
| Molecular Formula | C7H7ClN2O |
| Molecular Weight (g/mol) | 170.60 |
| MDL Number | MFCD00007603 |
| InChI Key | PKBGHORNUFQAAW-UHFFFAOYSA-N |
| Synonym | 4-chlorobenzhydrazide, 4-chlorobenzoylhydrazine, 4-chlorobenzoic hydrazide, p-chlorobenzohydrazide, 4-chlorobenzoic acid hydrazide, p-chlorobenzoylhydrazine, p-chlorobenzoic hydrazide, p-chlorobenzoyl hydrazide, 4-chlorobenzoyl hydrazide, p-chlorobenzoic acid, hydrazide |
| PubChem CID | 10816 |
| IUPAC Name | 4-chlorobenzohydrazide |
| SMILES | NNC(=O)C1=CC=C(Cl)C=C1 |
Chemical Identifiers
| 536-40-3 | |
| 170.60 | |
| PKBGHORNUFQAAW-UHFFFAOYSA-N | |
| 10816 | |
| NNC(=O)C1=CC=C(Cl)C=C1 |
| C7H7ClN2O | |
| MFCD00007603 | |
| 4-chlorobenzhydrazide, 4-chlorobenzoylhydrazine, 4-chlorobenzoic hydrazide, p-chlorobenzohydrazide, 4-chlorobenzoic acid hydrazide, p-chlorobenzoylhydrazine, p-chlorobenzoic hydrazide, p-chlorobenzoyl hydrazide, 4-chlorobenzoyl hydrazide, p-chlorobenzoic acid, hydrazide | |
| 4-chlorobenzohydrazide |
Specifications
| 536-40-3 | |
| White-Yellow | |
| MFCD00007603 | |
| 4-chlorobenzhydrazide, 4-chlorobenzoylhydrazine, 4-chlorobenzoic hydrazide, p-chlorobenzohydrazide, 4-chlorobenzoic acid hydrazide, p-chlorobenzoylhydrazine, p-chlorobenzoic hydrazide, p-chlorobenzoyl hydrazide, 4-chlorobenzoyl hydrazide, p-chlorobenzoic acid, hydrazide | |
| NNC(=O)C1=CC=C(Cl)C=C1 | |
| 170.60 | |
| 170.60 | |
| Crystalline Powder |
| 166°C | |
| C7H7ClN2O | |
| 5 g | |
| PKBGHORNUFQAAW-UHFFFAOYSA-N | |
| 4-chlorobenzohydrazide | |
| 10816 | |
| ≥98.0% (GC,T) | |
| 4-Chlorobenzohydrazide |
Safety and Handling
TSCA : No