missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzofuro[3,2-d]pyrimidine 98.0+%, TCI America™

Catalog No. C2800200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2800200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C2800200MG Supplier TCI America Supplier No. C2800200MG
Only null left

Chemical Identifiers

CAS 39876-88-5
Molecular Formula C10H5ClN2O
Molecular Weight (g/mol) 204.613
MDL Number MFCD00763943
InChI Key ABRHSRPOYMSBOI-UHFFFAOYSA-N
PubChem CID 609295
IUPAC Name 4-chloro-[1]benzofuro[3,2-d]pyrimidine
SMILES C1=CC=C2C(=C1)C3=C(O2)C(=NC=N3)Cl

Specifications

Melting Point 146°C
Color Yellow
Quantity 200 mg
Formula Weight 204.61
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chlorobenzofuro[3,2-d]pyrimidine