accessibility menu, dialog, popup

UserName

ClNNO

CAS RN 39876-88-5

IUPAC Name: 6-chloro-8-oxa-3,5-diazatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
Synonyms: 6-chloro-8-oxa-3,5-diazatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
Molecular Weight (g/mol): 204.61
Molecular Formula: C10H5ClN2O
InChi Key: ABRHSRPOYMSBOI-UHFFFAOYSA-N
SMILES: ClC1=NC=NC2=C1OC1=C2C=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results