missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chlorobenzhydryl Chloride 96.0+%, TCI America™
$75.08 - $587.18
Chemical Identifiers
| CAS | 134-83-8 |
|---|---|
| Molecular Formula | C13H10Cl2 |
| Molecular Weight (g/mol) | 237.123 |
| MDL Number | MFCD00000856 |
| InChI Key | ALKWTKGPKKAZMN-UHFFFAOYSA-N |
| Synonym | 4-chlorobenzhydryl chloride, 4-chlorobenzhydrylchloride, p-chlorobenzhydryl chloride, 1-chloro-4-chloro phenyl methyl benzene, benzene, 1-chloro-4-chlorophenylmethyl, chloro 4-chlorophenyl phenylmethane, chloro p-chlorophenyl phenylmethane, methane, chloro p-chlorophenyl phenyl, 1-chloro-4-chlorophenylmethyl benzene |
| PubChem CID | 241584 |
| IUPAC Name | 1-chloro-4-[chloro(phenyl)methyl]benzene |
| SMILES | C1=CC=C(C=C1)C(C2=CC=C(C=C2)Cl)Cl |
Chemical Identifiers
| 134-83-8 | |
| 237.123 | |
| ALKWTKGPKKAZMN-UHFFFAOYSA-N | |
| 241584 | |
| C1=CC=C(C=C1)C(C2=CC=C(C=C2)Cl)Cl |
| C13H10Cl2 | |
| MFCD00000856 | |
| 4-chlorobenzhydryl chloride, 4-chlorobenzhydrylchloride, p-chlorobenzhydryl chloride, 1-chloro-4-chloro phenyl methyl benzene, benzene, 1-chloro-4-chlorophenylmethyl, chloro 4-chlorophenyl phenylmethane, chloro p-chlorophenyl phenylmethane, methane, chloro p-chlorophenyl phenyl, 1-chloro-4-chlorophenylmethyl benzene | |
| 1-chloro-4-[chloro(phenyl)methyl]benzene |
Specifications
| 134-83-8 | |
| 160°C | |
| MFCD00000856 | |
| 1760 | |
| ALKWTKGPKKAZMN-UHFFFAOYSA-N | |
| 1-chloro-4-[chloro(phenyl)methyl]benzene | |
| 241584 | |
| ≥96.0% (GC) | |
| 4-Chlorobenzhydryl Chloride |
| Yellow | |
| C13H10Cl2 | |
| 25 g | |
| 4-chlorobenzhydryl chloride, 4-chlorobenzhydrylchloride, p-chlorobenzhydryl chloride, 1-chloro-4-chloro phenyl methyl benzene, benzene, 1-chloro-4-chlorophenylmethyl, chloro 4-chlorophenyl phenylmethane, chloro p-chlorophenyl phenylmethane, methane, chloro p-chlorophenyl phenyl, 1-chloro-4-chlorophenylmethyl benzene | |
| C1=CC=C(C=C1)C(C2=CC=C(C=C2)Cl)Cl | |
| 237.123 | |
| 237.12 | |
| Liquid |
Safety and Handling
EINECSNumber : (9)-0387
TSCA : Yes